Ethyl-2-(3-azetidinyloxy)benzoate

Supplier: Matrix Scientific

049431-500MG 049431-2.5G
101887-628EA 364.62 USD
101887-628 101887-630
Ethyl-2-(3-azetidinyloxy)benzoate
Ethyl-2-(3-azetidinyloxy)benzoate

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD13559900
CAS Number: 1220028-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR