3-Azetidinyl-5-chloro[1,1'-biphenyl]-2-yl ether

Supplier: Matrix Scientific

049430-500MG 049430-2.5G
101887-624EA 364.62 USD
101887-624 101887-626
3-Azetidinyl-5-chloro[1,1'-biphenyl]-2-yl ether
3-Azetidinyl-5-chloro[1,1'-biphenyl]-2-yl ether

Hazard: Irritant

Formula: C₁₅H₁₄ClNO
MDL Number: MFCD13559899
CAS Number: 1219948-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR