3-[2-(sec-Butyl)-4-chlorophenoxy]azetidine

Supplier: Matrix Scientific

049428-500MG 049428-2.5G
101887-616EA 364.62 USD
101887-616 101887-618
3-[2-(sec-Butyl)-4-chlorophenoxy]azetidine
3-[2-(sec-Butyl)-4-chlorophenoxy]azetidine

Hazard: Irritant

Formula: C₁₃H₁₈ClNO
MDL Number: MFCD13559897
CAS Number: 1220038-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR