3-[4-(tert-Butyl)-2-chlorophenoxy]azetidine

Supplier: Matrix Scientific

049427-500MG 049427-2.5G
101887-612EA 364.62 USD
101887-612 101887-614
3-[4-(tert-Butyl)-2-chlorophenoxy]azetidine
3-[4-(tert-Butyl)-2-chlorophenoxy]azetidine

Hazard: Irritant

Formula: C₁₃H₁₈ClNO
MDL Number: MFCD13559896
CAS Number: 1220027-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR