3-[2-(tert-Butyl)-4-chlorophenoxy]azetidine
Supplier: Matrix Scientific
049426-500MG
049426-2.5G
101887-608EA
364.62
USD
101887-608
101887-610
3-[2-(tert-Butyl)-4-chlorophenoxy]azetidine
3-[2-(tert-Butyl)-4-chlorophenoxy]azetidine
Hazard: Irritant
Formula:
C₁₃H₁₈ClNO |
MDL Number:
MFCD13559895 CAS Number: 1146960-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...