3-[2-Chloro-4-(tert-pentyl)phenoxy]azetidine

Supplier: Matrix Scientific

049425-500MG 049425-2.5G
101887-604EA 364.62 USD
101887-604 101887-606
3-[2-Chloro-4-(tert-pentyl)phenoxy]azetidine
3-[2-Chloro-4-(tert-pentyl)phenoxy]azetidine

Hazard: Irritant

Formula: C₁₄H₂₀ClNO
MDL Number: MFCD13559894
CAS Number: 1220021-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR