3-Azetidinyl-2-bromo-4-(tert-pentyl)phenyl ether

Supplier: Matrix Scientific

049424-500MG 049424-2.5G
101887-600EA 364.62 USD
101887-600 101887-602
3-Azetidinyl-2-bromo-4-(tert-pentyl)phenyl ether
3-Azetidinyl-2-bromo-4-(tert-pentyl)phenyl ether

Hazard: Irritant

Formula: C₁₄H₂₀BrNO
MDL Number: MFCD13559893
CAS Number: 1219976-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR