3-Azetidinyl-2-chloro-4-methoxyphenyl ether

Supplier: Matrix Scientific

049422-500MG 049422-2.5G
101887-592EA 364.62 USD
101887-592 101887-594
3-Azetidinyl-2-chloro-4-methoxyphenyl ether
3-Azetidinyl-2-chloro-4-methoxyphenyl ether

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₂
MDL Number: MFCD13559891
CAS Number: 1220038-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR