3-(2-Chloro-4-methylphenoxy)azetidine

Supplier: Matrix Scientific

049419-500MG 049419-2.5G
101887-580EA 364.62 USD
101887-580 101887-582
3-(2-Chloro-4-methylphenoxy)azetidine
3-(2-Chloro-4-methylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₂ClNO
MDL Number: MFCD13559888
CAS Number: 1219976-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR