3-[4-(tert-Butyl)-2-methylphenoxy]azetidine

Supplier: Matrix Scientific

049414-500MG 049414-2.5G
101887-560EA 364.62 USD
101887-560 101887-562
3-[4-(tert-Butyl)-2-methylphenoxy]azetidine
3-[4-(tert-Butyl)-2-methylphenoxy]azetidine

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD13559883
CAS Number: 1220027-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR