3-Azetidinyl-5-bromo[1,1'-biphenyl]-2-yl ether hydrochloride

Supplier: Matrix Scientific

049413-500MG 049413-2.5G
101887-556EA 364.62 USD
101887-556 101887-558
3-Azetidinyl-5-bromo[1,1'-biphenyl]-2-yl ether hydrochloride
3-Azetidinyl-5-bromo[1,1'-biphenyl]-2-yl ether hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₅BrClNO
MDL Number: MFCD20526327
CAS Number: 1354550-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR