3-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]azetidine

Supplier: Matrix Scientific

049412-500MG 049412-2.5G
101887-552EA 364.62 USD
101887-552 101887-554
3-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]azetidine
3-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]azetidine

Hazard: Irritant

Formula: C₁₈H₂₀BrNO
MW: 346.28 g/mol
MDL Number: MFCD13559881
CAS Number: 1219981-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR