3-(4-Chloro-2-methylphenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049411-500MG 049411-2.5G
101887-548EA 364.62 USD
101887-548 101887-550
3-(4-Chloro-2-methylphenoxy)azetidine hydrochloride
3-(4-Chloro-2-methylphenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃Cl₂NO
MDL Number: MFCD26936319
CAS Number: 954223-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR