3-(4-Bromo-3-methylphenoxy)azetidine

Supplier: Matrix Scientific

049410-500MG 049410-2.5G
101887-544EA 364.62 USD
101887-544 101887-546
3-(4-Bromo-3-methylphenoxy)azetidine
3-(4-Bromo-3-methylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₂BrNO
MDL Number: MFCD13559880
CAS Number: 1220038-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR