3-[2-Bromo-4-(tert-butyl)phenoxy]azetidine

Supplier: Matrix Scientific

049408-500MG 049408-2.5G
101887-536EA 364.62 USD
101887-536 101887-538
3-[2-Bromo-4-(tert-butyl)phenoxy]azetidine
3-[2-Bromo-4-(tert-butyl)phenoxy]azetidine

Hazard: Irritant

Formula: C₁₃H₁₈BrNO
MDL Number: MFCD13559878
CAS Number: 1219948-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR