3-Azetidinyl-3-bromo[1,1'-biphenyl]-4-yl ether

Supplier: Matrix Scientific

049407-500MG 049407-2.5G
101887-532EA 364.62 USD
101887-532 101887-534
3-Azetidinyl-3-bromo[1,1'-biphenyl]-4-yl ether
3-Azetidinyl-3-bromo[1,1'-biphenyl]-4-yl ether

Hazard: Irritant

Formula: C₁₅H₁₄BrNO
MDL Number: MFCD13559877
CAS Number: 1219960-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR