3-[2-Bromo-4-(sec-butyl)phenoxy]azetidine
Supplier: Matrix Scientific
049402-500MG
049402-2.5G
101887-512EA
364.62
USD
101887-512
101887-514
3-[2-Bromo-4-(sec-butyl)phenoxy]azetidine
3-[2-Bromo-4-(sec-butyl)phenoxy]azetidine
Hazard: Irritant
Formula:
C₁₃H₁₈BrNO |
MDL Number:
MFCD13559872 CAS Number: 1219956-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...