3-[(1,6-Dibromo-2-naphthyl)oxy]azetidine

Supplier: Matrix Scientific

049401-500MG 049401-2.5G
101887-508EA 364.62 USD
101887-508 101887-510
3-[(1,6-Dibromo-2-naphthyl)oxy]azetidine
3-[(1,6-Dibromo-2-naphthyl)oxy]azetidine

Hazard: Irritant

Formula: C₁₃H₁₁Br₂NO
MDL Number: MFCD13559871
CAS Number: 1219976-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR