3-[(2,4-Dichloro-1-naphthyl)oxy]azetidine

Supplier: Matrix Scientific

049400-500MG 049400-2.5G
101887-504EA 364.62 USD
101887-504 101887-506
3-[(2,4-Dichloro-1-naphthyl)oxy]azetidine
3-[(2,4-Dichloro-1-naphthyl)oxy]azetidine

Hazard: Irritant

Formula: C₁₃H₁₁Cl₂NO
MDL Number: MFCD13559870
CAS Number: 719993-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR