3-(2-Bromo-4-chloro-3,5-dimethylphenoxy)azetidine
Supplier: Matrix Scientific
049398-500MG
049398-2.5G
101887-496EA
364.62
USD
101887-496
101887-498
3-(2-Bromo-4-chloro-3,5-dimethylphenoxy)azetidine
3-(2-Bromo-4-chloro-3,5-dimethylphenoxy)azetidine
Hazard: Irritant
Formula:
C₁₁H₁₃BrClNO |
MDL Number:
MFCD13559868 CAS Number: 1220027-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...