3-(4-Benzyl-2-bromophenoxy)azetidine

Supplier: Matrix Scientific

049397-500MG 049397-2.5G
101887-492EA 364.62 USD
101887-492 101887-494
3-(4-Benzyl-2-bromophenoxy)azetidine
3-(4-Benzyl-2-bromophenoxy)azetidine

Hazard: Irritant

Formula: C₁₆H₁₆BrNO
MW: 318.22 g/mol
MDL Number: MFCD13559867
CAS Number: 1220027-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR