3-(2-Bromo-4-methoxyphenoxy)azetidine

Supplier: Matrix Scientific

049395-500MG 049395-2.5G
101887-484EA 364.62 USD
101887-484 101887-486
3-(2-Bromo-4-methoxyphenoxy)azetidine
3-(2-Bromo-4-methoxyphenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₂BrNO₂
MDL Number: MFCD13559865
CAS Number: 1220027-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR