2-[Methyl(3-pyridinylcarbonyl)amino]acetic acid
Supplier: Matrix Scientific
049394-500MG
049394-2.5G
101887-480EA
364.62
USD
101887-480
101887-482
2-[Methyl(3-pyridinylcarbonyl)amino]acetic acid
2-[Methyl(3-pyridinylcarbonyl)amino]acetic acid
Hazard: Irritant
Formula:
C₉H₁₀N₂O₃ |
MDL Number:
MFCD09050995 CAS Number: 926217-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...