2-Methyl-N-(3-pyridinylcarbonyl)alanine

Supplier: Matrix Scientific

049393-500MG 049393-2.5G
101887-476EA 364.62 USD
101887-476 101887-478
2-Methyl-N-(3-pyridinylcarbonyl)alanine
2-Methyl-N-(3-pyridinylcarbonyl)alanine

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃
MW: 208.22 g/mol
MDL Number: MFCD13559864
CAS Number: 1220027-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR