1-(3-Pyridinylcarbonyl)-4-piperidinecarboxylic acid

Supplier: Matrix Scientific

049392-500MG 049392-2.5G
101887-472EA 364.62 USD
101887-472 101887-474
1-(3-Pyridinylcarbonyl)-4-piperidinecarboxylic acid
1-(3-Pyridinylcarbonyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃
MDL Number: MFCD00434437
CAS Number: 67691-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR