1-(3-Pyridinylcarbonyl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

049391-500MG 049391-2.5G
101887-468EA 364.62 USD
101887-468 101887-470
1-(3-Pyridinylcarbonyl)-3-piperidinecarboxylic acid
1-(3-Pyridinylcarbonyl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃
MDL Number: MFCD09046811
CAS Number: 67691-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR