1-(3-Pyridinylcarbonyl)proline

Supplier: Matrix Scientific

049389-500MG 049389-2.5G
101887-460EA 364.62 USD
101887-460 101887-462
1-(3-Pyridinylcarbonyl)proline
1-(3-Pyridinylcarbonyl)proline

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD00448257
CAS Number: 1101194-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR