N-Nicotinoylglycine
Supplier: Matrix Scientific
Synonyms:
2-(Nicotinamido)acetic acid, Nicotinuric Acid
049387-500MG
049387-2.5G
101887-452EA
364.62
USD
101887-452
101887-454
N-Nicotinoylglycine
N-Nicotinoylglycine
Hazard: Irritant
Formula:
C₈H₈N₂O₃ MW: 180.16 g/mol |
MDL Number:
MFCD00023578 CAS Number: 583-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...