3-Pyrrolidinyl nicotinate hydrochloride

Supplier: Matrix Scientific

049384-500MG 049384-2.5G
101887-440EA 364.62 USD
101887-440 101887-442
3-Pyrrolidinyl nicotinate hydrochloride
3-Pyrrolidinyl nicotinate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MDL Number: MFCD13559861
CAS Number: 1220038-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR