3-Pyrrolidinylmethyl nicotinate hydrochloride
Supplier: Matrix Scientific
049383-500MG
049383-2.5G
101887-436EA
364.62
USD
101887-436
101887-438
3-Pyrrolidinylmethyl nicotinate hydrochloride
3-Pyrrolidinylmethyl nicotinate hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅ClN₂O₂ |
MDL Number:
MFCD13559860 CAS Number: 1220021-63-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...