2-(2-Piperidinyl)ethyl nicotinate hydrochloride

Supplier: Matrix Scientific

049382-500MG 049382-2.5G
101887-432EA 364.62 USD
101887-432 101887-434
2-(2-Piperidinyl)ethyl nicotinate hydrochloride
2-(2-Piperidinyl)ethyl nicotinate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂
MDL Number: MFCD13559859
CAS Number: 1220021-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR