2-(3-Piperidinyl)ethyl nicotinate hydrochloride

Supplier: Matrix Scientific

049381-500MG 049381-2.5G
101887-428EA 364.62 USD
101887-428 101887-430
2-(3-Piperidinyl)ethyl nicotinate hydrochloride
2-(3-Piperidinyl)ethyl nicotinate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂
MDL Number: MFCD13559858
CAS Number: 1219976-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR