2-(4-Piperidinyl)ethyl nicotinate hydrochloride

Supplier: Matrix Scientific

049380-500MG 049380-2.5G
101887-424EA 364.62 USD
101887-424 101887-426
2-(4-Piperidinyl)ethyl nicotinate hydrochloride
2-(4-Piperidinyl)ethyl nicotinate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂
MW: 270.76 g/mol
MDL Number: MFCD13559857
CAS Number: 1219981-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR