3-Piperidinylmethyl nicotinate hydrochloride

Supplier: Matrix Scientific

049378-500MG 049378-2.5G
101887-416EA 364.62 USD
101887-416 101887-418
3-Piperidinylmethyl nicotinate hydrochloride
3-Piperidinylmethyl nicotinate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂
MW: 256.73 g/mol
MDL Number: MFCD13559855
CAS Number: 1219976-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR