3-Azetidinyl nicotinate

Supplier: Matrix Scientific

049377-500MG 049377-2.5G
101887-412EA 364.62 USD
101887-412 101887-414
3-Azetidinyl nicotinate
3-Azetidinyl nicotinate

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
MDL Number: MFCD13559854
CAS Number: 1220021-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR