4-Piperidinyl nicotinate hydrochloride
Supplier: Matrix Scientific
049376-500MG
049376-2.5G
101887-408EA
364.62
USD
101887-408
101887-410
4-Piperidinyl nicotinate hydrochloride
4-Piperidinyl nicotinate hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅ClN₂O₂ |
MDL Number:
MFCD13559853 CAS Number: 1219948-64-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...