3-Piperidinyl nicotinate hydrochloride

Supplier: Matrix Scientific

049375-500MG 049375-2.5G
101887-404EA 364.62 USD
101887-404 101887-406
3-Piperidinyl nicotinate hydrochloride
3-Piperidinyl nicotinate hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₂
MDL Number: MFCD13559852
CAS Number: 1219981-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR