Benzoyl-DL-alanine

Supplier: Matrix Scientific

049370-500MG 049370-2.5G
101887-384EA 364.62 USD
101887-384 101887-386
Benzoyl-DL-alanine
Benzoyl-DL-alanine

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
MDL Number: MFCD00020393
CAS Number: 1205-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR