3-Pyrrolidinyl benzoate hydrochloride

Supplier: Matrix Scientific

049367-500MG 049367-2.5G
101887-372EA 364.62 USD
101887-372 101887-374
3-Pyrrolidinyl benzoate hydrochloride
3-Pyrrolidinyl benzoate hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₂
MDL Number: MFCD13559849
CAS Number: 99855-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR