2-(4-Piperidinyl)ethyl benzoate hydrochloride
Supplier: Matrix Scientific
049363-500MG
049363-2.5G
101887-356EA
166.15
USD
101887-356
101887-358
2-(4-Piperidinyl)ethyl benzoate hydrochloride
2-(4-Piperidinyl)ethyl benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₀ClNO₂ |
MDL Number:
MFCD13559846 CAS Number: 1220021-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...