4-Piperidinylmethyl benzoate hydrochloride

Supplier: Matrix Scientific

049362-500MG 049362-2.5G
101887-352EA 364.62 USD
101887-352 101887-354
4-Piperidinylmethyl benzoate hydrochloride
4-Piperidinylmethyl benzoate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₂
MDL Number: MFCD13559845
CAS Number: 1220021-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR