3-Piperidinylmethyl benzoate hydrochloride
Supplier: Matrix Scientific
049361-500MG
049361-2.5G
101887-348EA
166.15
USD
101887-348
101887-350
3-Piperidinylmethyl benzoate hydrochloride
3-Piperidinylmethyl benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈ClNO₂ |
MDL Number:
MFCD13559844 CAS Number: 1219972-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...