3-Azetidinyl benzoate

Supplier: Matrix Scientific

049360-500MG 049360-2.5G
101887-344EA 364.62 USD
101887-344 101887-346
3-Azetidinyl benzoate
3-Azetidinyl benzoate

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD13559843
CAS Number: 1219980-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR