4-Piperidinyl benzoate hydrochloride
Supplier: Matrix Scientific
049359-500MG
049359-2.5G
101887-340EA
364.62
USD
101887-340
101887-342
4-Piperidinyl benzoate hydrochloride
4-Piperidinyl benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₆ClNO₂ |
MDL Number:
MFCD13559842 CAS Number: 67098-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...