N-(Cyclopentylcarbonyl)-β-alanine
Supplier: Matrix Scientific
049357-500MG
049357-2.5G
101887-332EA
364.62
USD
101887-332
101887-334
N-(Cyclopentylcarbonyl)-β-alanine
N-(Cyclopentylcarbonyl)-beta-alanine
Hazard: Irritant
Formula:
C₉H₁₅NO₃ MW: 185.22 g/mol |
MDL Number:
MFCD08445014 CAS Number: 923225-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...