2-[(Cyclopentylcarbonyl)(methyl)amino]acetic acid

Supplier: Matrix Scientific

049356-500MG 049356-2.5G
101887-328EA 592.31 USD
101887-328 101887-330
2-[(Cyclopentylcarbonyl)(methyl)amino]acetic acid
2-[(Cyclopentylcarbonyl)(methyl)amino]acetic acid

Hazard: Irritant

Formula: C₉H₁₅NO₃
MDL Number: MFCD09048941
CAS Number: 926252-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR