1-(Cyclopentylcarbonyl)-4-piperidinecarboxylic acid

Supplier: Matrix Scientific

049354-500MG 049354-2.5G
101887-320EA 364.62 USD
101887-320 101887-322
1-(Cyclopentylcarbonyl)-4-piperidinecarboxylic acid
1-(Cyclopentylcarbonyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₉NO₃
MDL Number: MFCD08443273
CAS Number: 147636-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR