1-(Cyclopentylcarbonyl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

049353-500MG 049353-2.5G
101887-316EA 166.15 USD
101887-316 101887-318
1-(Cyclopentylcarbonyl)-3-piperidinecarboxylic acid
1-(Cyclopentylcarbonyl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₉NO₃
MDL Number: MFCD08442439
CAS Number: 926227-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR