1-(Cyclopentylcarbonyl)-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

049352-500MG 049352-2.5G
101887-312EA 364.62 USD
101887-312 101887-314
1-(Cyclopentylcarbonyl)-2-piperidinecarboxylic acid
1-(Cyclopentylcarbonyl)-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₉NO₃
MDL Number: MFCD08445234
CAS Number: 1044637-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR