1-(Cyclopentylcarbonyl)proline

Supplier: Matrix Scientific

049351-500MG 049351-2.5G
101887-308EA 364.62 USD
101887-308 101887-310
1-(Cyclopentylcarbonyl)proline
1-(Cyclopentylcarbonyl)proline

Hazard: Irritant

Formula: C₁₁H₁₇NO₃
MDL Number: MFCD00522584
CAS Number: 1417985-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR